Citrate, 0.5M buffer soln., pH 4.5
Citrate, 0.5M buffer soln., pH 4.5
Citrate, 0.5M buffer soln., pH 4.5
Thermo Scientific Chemicals

Citrate, 0.5M buffer soln., pH 4.5

CAS: 6132-04-3294.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60024.AK
also known as J60024-AK
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 30-Jun-2026
107.00
Save 16.35
Each
Specifications
CAS4-3-6132
ColorColorless
OdorOdorless
Recommended StorageKeep cold
Concentration0.5M buffer soln.
View more
Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citric acid is used as a substrate for citrate lyase, a buffer component, an anticoagulant. For anticoagulation use it is typically used at a concentration of approximately 0.129 M.

Solubility
Fully miscible with water. Insoluble in ethanol.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. H.U. Bergmeyer. Substrate for citrate lyase. ed. Methods of Enzymatic Analysis 3rd ed., 1983, 2173.
  2. D M Ramsay; E P Robertson; E MacArthur. Comparison of anticoagulants for the preservation of prothrombin time specimens. Journal of Clinical Pathology, 1977, 30(8), 766-769.