Elastase Substrate V
Elastase Substrate V
Elastase Substrate V
Thermo Scientific Chemicals

Elastase Substrate V

627.70 g/mol
Have Questions?
Catalog NumberQuantity
J60093.MB
also known as J60093-MB
25 mg
Catalog number J60093.MB
also known as J60093-MB
Price (USD)
258.65
Special offer
Online exclusive
Ends: 30-Jun-2026
304.00
Save 45.35 (15%)
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
258.65
Special offer
Online exclusive
Ends: 30-Jun-2026
304.00
Save 45.35 (15%)
Each
Specifications
ColorWhite
For Use With (Application)Fluorogenic substrate for quantification of human leukocyte and porcine pancreatic elastase
Molecular Weight (g/mol)627.70
Quantity25 mg
Recommended StorageStore at -20°C
View more
Fluorogenic substrate for quantification of human leukocyte and porcine pancreatic elastase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only