Aphidicolin
Aphidicolin
Aphidicolin
Aphidicolin
Thermo Scientific™

Aphidicolin

A DNA polymerase inhibitor, blocks the cell cycle at the early S-phase | CAS: 38966-21-1 | C20H34O4, C20H34O4 | 338.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number J60236.LB0
also known as J60236-#0
Price (USD)
574.65
Special offer
Online exclusive
Ends: 30-Jun-2026
676.00
Save 101.35 (15%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
574.65
Special offer
Online exclusive
Ends: 30-Jun-2026
676.00
Save 101.35 (15%)
Each
Chemical Identifiers
CAS38966-21-1
IUPAC Name(1S,2S,5R,6R,7R,10S,12R,13R)-6,13-bis(hydroxymethyl)-2,6-dimethyltetracyclo[10.3.1.0¹,¹⁰.0²,⁷]hexadecane-5,13-diol
Molecular FormulaC20H34O4
InChI KeyNOFOAYPPHIUXJR-APNQCZIXSA-N
SMILESC[C@@]1(CO)[C@H](O)CC[C@@]2(C)[C@H]1CC[C@H]1C[C@@H]3C[C@]21CC[C@]3(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
Purity≥97.0%
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C