Tioconazole, 98+%
Thermo Scientific Chemicals

Tioconazole, 98+%

Potent inhibitor of cytochrome-P450 | CAS: 65899-73-2 | C16H13Cl3N2OS
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60459.03
also known as J60459-03
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Specifications
Assay≥98%
CAS65899-73-2
DescriptionWhite to Off-white
EINECS Number265-973-8
For Use With (Application)Potent inhibitor of cytochrome-P450
View more
Potent inhibitor of cytochrome-P450

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only