N-Acetyl-D-sphingosine, 98%
N-Acetyl-D-sphingosine, 98%
N-Acetyl-D-sphingosine, 98%
N-Acetyl-D-sphingosine, 98%
Thermo Scientific Chemicals

N-Acetyl-D-sphingosine, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J60534.MA
also known as J60534-MA
Price (USD)
173.65
Special offer
Online exclusive
Ends: 30-Jun-2026
204.00
Save 30.35 (15%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
173.65
Special offer
Online exclusive
Ends: 30-Jun-2026
204.00
Save 30.35 (15%)
Each
Specifications
Assay98%
CAS3102-57-6
DescriptionWhite
For Use With (Application)Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases
FormPowder
View more
Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells
  • Activates protein phosphatases
RUO – Research Use Only
Contents & Storage
-30°C to -10°C