This product is animal origin and can only be shipped within the United States
Hyaluronic acid, bovine vitreous humor
Hyaluronic acid, bovine vitreous humor
Hyaluronic acid, bovine vitreous humor
Thermo Scientific Chemicals

Hyaluronic acid, bovine vitreous humor

Hyaluronic acid from bovine vitreous humor, CAS # 9004-61-9, is a naturally derived glycosaminoglycan that shows an angiogenic activity that promotes wound healing processes. | CAS: 9004-61-9 | C28H44N2O23 | 776.651 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60566.MF
also known as J60566-MF
Price (USD)
580.65
Online Exclusive
645.00
Save 64.35 (10%)
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
580.65
Online Exclusive
645.00
Save 64.35 (10%)
Each
Chemical Identifiers
CAS9004-61-9
IUPAC Namesodium (2S,3S,4R,5R,6R)-3-{[(2S,3R,4R,5S,6R)-4-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3-acetamido-5-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-6-{[(2R,3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4,5-dihydroxyoxane-2-carboxylic acid
Molecular FormulaC28H44N2NaO23
InChI KeyYWIVKILSMZOHHF-QJZPQSOGSA-N
SMILES[Na+].CC(=O)N[C@H]1[C@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O[C@@H]1O[C@@H]([C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)[C@H]2NC(C)=O)[C@H](O)[C@H]1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
CommentSubstrate suitability: Satisfactory for hyaluronidase assay
FormPowder
Hyaluronic acid is a naturally derived, non - immunogenic, non - adhesive glycosaminoglycan that plays a prominent role in various wound - healing processes, as it as it is naturally angiogenic when degraded to small fragments. Hyaluronic acid promotes early inflammation which is critical for initiating wound healing, but then moderates later stages of the process, allowing matrix stabilization and reduction of long term inflammation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hyaluronic acid, derived from bovine vitreous humor, is a naturally derived, non-immunogenic, non-adhesive glycosaminoglycan that shows an angiogenic activity promoting wound-healing processes when degraded to small fragments

Application

  • It is considered a mucopolysaccharide composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine, which forms the core of complex proteoglycan aggregates found in the extracellular matrix
  • It has been used to study the activation of the MAPK/ERK pathway following infection of macrophages with Leishmania amazonensis
RUO – Research Use Only

General References:

  1. Yuushi Okumura. Serase-1B, a new splice variant of polyserase-1/TMPRSS9, activates urokinase-type plasminogen activator and the proteolytic activation is negatively regulated by glycosaminoglycans. Biochemical Journal. 2006, 400, (4), 551-555.