Thermo Scientific Chemicals
Ingenol 3-angelate, 98%
Catalog number: J60601.MCR
1 mg, Each
Thermo Scientific Chemicals
Ingenol 3-angelate, 98%
Catalog number: J60601.MCR
1 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60601.MCR
also known as J60601-M^
Price (USD)
Price: 316.00
Online price: 275.65
Your price:
Quantity
-
Description
Ingenol 3-Angelate is used as a PKC activator, anticancer, and immunostimulant compound. It is also used to exhibits antileukemic activity, induces nuclear translocation of PKCδ, and induces apoptosis in acute myeloid leukemia (AML) cell lines and primary AML blast cells. Also displays antiproliferative effects and induces apoptosis in Colo205 cells.