Ionomycin Calcium Salt, 99%
Ionomycin Calcium Salt, 99%
Ionomycin Calcium Salt, 99%
Thermo Scientific Chemicals

Ionomycin Calcium Salt, 99%

CAS: 56092-82-1 | C41H70CaO9, C41H70CaO9 | 747.08 g/mol
Have Questions?
Catalog NumberQuantity
J60628.LB0
also known as J60628-#0
5 mg
Catalog number J60628.LB0
also known as J60628-#0
Price (USD)
203.65
Online Exclusive
226.00
Save 22.35 (10%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
203.65
Online Exclusive
226.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS56092-82-1
IUPAC Namecalcium (4R,6S,8S,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,2'R,5S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9,11-dioxodocos-16-en-10-idoate
Molecular FormulaC41H70CaO9
InChI KeyRULWQOBWZOQCOY-SQNLDGQHSA-M
SMILES[Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Purity≥98.5%
Ionomycin calcium salt is used as a polyether antibiotic and apoptotic degeneration inducer. Ionomycin induces apoptotic degeneration of embryonic cortical neurons and cell cycle arrest at G1 phase. It is more effective than A23187 (sc-3591) and provides a useful tool for increasing intracellular Ca2+ concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C