This product is animal origin and can only be shipped within the United States
Lysozyme, chicken egg white
Lysozyme, chicken egg white
Lysozyme, chicken egg white
Thermo Scientific Chemicals

Lysozyme, chicken egg white

Lysozyme, chicken egg white, CAS # 12650-88-3, is an enzyme that catalyzes the hydrolysis of peptidoglycans found in gram-positive bacterial cell walls, causing bacterial cell lysis. | CAS: 12650-88-3 | C125H196N40O36S2
Have Questions?
Change viewbuttonViewtableView
Quantity:
1g
25g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60701.22
also known as J60701-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS12650-88-3
IUPAC Name4-({1-[(1-{[1-({1-[(1-{[5-amino-1-({4-carbamimidamido-1-[(1-{[({1-[(1-{[1-({1-[(4-carbamimidamido-1-{[({1-[(1-{[1-({[(2-carbamoyl-1-carboxyethyl)carbamoyl]methyl}carbamoyl)-3-methylbutyl]carbamoyl}-2-hydroxyethyl)carbamoyl]-2-(4-hydroxyphenyl)ethyl}carbamoyl)methyl]carbamoyl}butyl)carbamoyl]-2-(4-hydroxyphenyl)ethyl}carbamoyl)-2-carbamoylethyl]carbamoyl}-2-carboxyethyl)carbamoyl]-3-methylbutyl}carbamoyl)methyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl)carbamoyl]butyl}carbamoyl)pentyl]carbamoyl}-3-(methylsulfanyl)propyl)carbamoyl]ethyl}carbamoyl)ethyl]carbamoyl}ethyl)carbamoyl]-3-methylbutyl}carbamoyl)-4-[2-(2-{2-[2-(2-amino-3-methylbutanamido)-3-phenylpropanamido]acetamido}-5-carbamimidamidopentanamido)-3-sulfanylpropanamido]butanoic acid
Molecular FormulaC125H196N40O36S2
InChI KeyJFXJPYIEDZSWNF-UHFFFAOYNA-N
SMILESCSCCC(NC(=O)C(C)NC(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)NC(=O)C(CCC(O)=O)NC(=O)C(CS)NC(=O)C(CCCNC(N)=N)NC(=O)CNC(=O)C(CC1=CC=CC=C1)NC(=O)C(N)C(C)C)C(=O)NC(CCCCN)C(=O)NC(CCCNC(N)=N)C(=O)NC(CC1=CN=CN1)C(=O)NCC(=O)NC(CC(C)C)C(=O)NC(CC(O)=O)C(=O)NC(CC(N)=O)C(=O)NC(CC1=CC=C(O)C=C1)C(=O)NC(CCCNC(N)=N)C(=O)NCC(=O)NC(CC1=CC=C(O)C=C1)C(=O)NC(CO)C(=O)NC(CC(C)C)C(=O)NCC(=O)NC(CC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
units/mgdw≥ 20000
Catalyzes the hydrolysis of peptidoglycans found in bacterial cell walls. Lysozyme catalyzes is utilized for the hydrolysis of peptidoglycans found in bacterial cell walls. It is used as a precursor in the production of spheroplasts. It is also used as a preservative against many food spoiling microorganisms. It is used in skin care in order to cure and prevent acne and bed sores, dental and oral conditions. It is used for lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific antigen. Further, it is used in non-pasteurized beer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Lysozyme, chicken egg white is a naturally occurring enzyme that hydrolyzes the peptidoglycans in the bacterial cell wall, resulting in cell wall lysis

Application

• Lysozyme is utilized for the hydrolysis of peptidoglycans found in bacterial cell walls
• It is used as a precursor in the production of spheroplasts in laboratory settings
• It is used for lysing E. coli and Streptomycetes for extraction purposes, such as extracting group-specific antigen

RUO – Research Use Only

General References:

  1. Bayramoglu, G.; Ozalp, V. C.; Yilmaz, M.; Guler, U.; Salih, B.; Arica, M. Y. Lysozyme specific aptamer immobilized MCM-41 silicate for single-step purification and quartz crystal microbalance (QCM)-based determination of lysozyme from chicken egg white. Microporous Mesoporous Mater. 2015, 207, 95-104.
  2. Gao, R.; Zhang, L.; Hao, Y.; Cui, X.; Liu, D.; Zhang, M.; Tang, Y. Novel polydopamine imprinting layers coated magnetic carbon nanotubes for specific separation of lysozyme from egg white. Talanta 2015, 144, 1125-1132.
Contents & Storage
Store at -20°C