2'-Deoxyguanosine monohydrate, 98+%
2'-Deoxyguanosine monohydrate, 98+%
2'-Deoxyguanosine monohydrate, 98+%
Thermo Scientific Chemicals

2'-Deoxyguanosine monohydrate, 98+%

CAS: 961-07-9 | C10H13N5O4 | 267.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number J60741.03
also known as J60741-03
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.70
Save 13.05 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.70
Save 13.05 (15%)
Each
Chemical Identifiers
CAS961-07-9
IUPAC Name2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular FormulaC10H13N5O4
InChI KeyYKBGVTZYEHREMT-DWFKHXNSNA-N
SMILESNC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms to standard
Appearance (Color)White to off-white
Assay (HPLC)≥98.0%
Water Content (Karl Fischer Titration)8.0% max
FormCrystalline powder
Deoxyguanosine (dG) is a purine nucleoside that upon sequential phosphoylation (kinases) forms dGTP which is used by DNA polymerases and reverse transcriptases to synthesis DNA(s). Deoxyguanosine is the most electron rich of the four canonical bases and includes many nucleophilic sites which are susceptible to oxidative damage. This makes deoxyguanosine and its oxidized derivatives useful reagents to study mechanisms of oxidative damage to nucleosides and nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxyguanosine (dG) is a purine nucleoside that upon sequential phosphoylation (kinases) forms dGTP which is used by DNA polymerases and reverse transcriptases to synthesis DNA(s). Deoxyguanosine is the most electron rich of the four canonical bases and includes many nucleophilic sites which are susceptible to oxidative damage. This makes deoxyguanosine and its oxidized derivatives useful reagents to study mechanisms of oxidative damage to nucleosides and nucleotides.

Solubility
Soluble in NH4OH 1 M: 50 mg/mL, clear to very faintly turbid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kasper Broedbaek et. al. Urinary 8-oxo-7,8-dihydro-2'-deoxyguanosine as a biomarker in type 2 diabetes. Free Radical Biology and Medicine. 2011, 51 (8), 1473-1479.
  2. Aaron M Fleming et. al. Copper/H2O2-mediated oxidation of 2'-deoxyguanosine in the presence of 2-naphthol leads to the formation of two distinct isomeric adducts.. Journal of Organic Chemistry. 2011, 76 (19), 7953-7963.