D-Glutamine, 99+%
D-Glutamine, 99+%
D-Glutamine, 99+%
Thermo Scientific Chemicals

D-Glutamine, 99+%

Kinase inhibitor; has antivasospastic properties | CAS: 5959-95-5 | C5H10N2O3 | 146.146 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number J60784.14
also known as J60784-14
Price (USD)
672.65
Online Exclusive
747.00
Save 74.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
672.65
Online Exclusive
747.00
Save 74.35 (10%)
Each
Chemical Identifiers
CAS5959-95-5
IUPAC Name(2R)-2-amino-4-carbamoylbutanoic acid
Molecular FormulaC5H10N2O3
InChI KeyZDXPYRJPNDTMRX-GSVOUGTGSA-N
SMILESN[C@H](CCC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥99.0%
Kinase inhibitor; has antivasospastic properties

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only