SSC buffer (20X)
Thermo Scientific Chemicals
SSC buffer (20X)
Suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2 L
4 L
Catalog number J60839.K3
also known as J60839-K3
Price (USD)
94.70
Each
Quantity:
2 L
Request bulk or custom format
Price (USD)
94.70
Each
Specifications
Recommended StorageAmbient temperatures
Physical FormLiquid
Quantity2 L
Unit SizeEach
SSC buffer is suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.