Adenosine-5'-triphosphate disodium salt hydrate, 98%
Adenosine-5'-triphosphate disodium salt hydrate, 98%
Adenosine-5'-triphosphate disodium salt hydrate, 98%
Thermo Scientific Chemicals

Adenosine-5'-triphosphate disodium salt hydrate, 98%

CAS: 34369-07-8 | C10H14N5Na2O13P3 | 551.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Catalog number J61125.09
also known as J61125-09
Price (USD)
172.65
Online Exclusive
192.00
Save 19.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
172.65
Online Exclusive
192.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS34369-07-8
IUPAC Namedisodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC10H14N5Na2O13P3
InChI KeyTTWYZDPBDWHJOR-WCYUCLFNNA-L
SMILES[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms to structure
Water Content (Karl Fischer Titration)≤10%
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay from Supplier's CofA≥98.0% (HPLC)
View more
Adenosine-5'-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

Solubility
Soluble in water (110 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), DMF (sparingly), and PBS pH 7.2 (∼10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Gordon. Extracellular ATP: effects, sources and fate. Biochem.J. 1986, 233,(2), 309-19.
  2. Guilbert et al. Dependence of P2-nucleotide receptor agonist-mediated endothelium-indepdent relaxation on ectonucleotidase acticity and A2A-receptors in rat portal vein. Br.J.Pharm. 1998, 123,(2), 1732-35.

Frequently asked questions (FAQs)

How do I make up a 20 mM solution of Adenosine-5'-triphosphate disodium salt hydrate, 98% (Cat. No. J61125.14)?

To make a 20 mM solution of Adenosine-5'-triphosphate disodium salt hydrate, 98% (Cat. No. J61125.14) for assays:

1. Calculate the amount of Adenosine-5'-triphosphate disodium salt hydrate, 98% needed:
- Molecular Weight: 551.15 g/mol
- Desired concentration: 20 mM
- Assuming that the volume of the solution is 1 L, the concentration is 0.02 mol/L

2. Amount needed (in grams) = Concentration (mol/L) x Molecular Weight (g/mol) x Volume (L)
- Amount needed = 0.02 mol/L x 551.15 g/mol x 1 L = 11.023 g

3. Dissolve the calculated amount in water:
- Dissolve 11.023 g of Adenosine-5'-triphosphate disodium salt hydrate, 98% in 1 L of water.

4. Adjust the volume if necessary:
- If you need a different volume, adjust the amount of Adenosine-5'-triphosphate disodium salt hydrate, 98% proportionally.

Note: Ensure the solution is clear and colorless to pale yellow, as per the solubility information provided.