Laemmli SDS sample buffer, reducing (6X)
Thermo Scientific Chemicals

Laemmli SDS sample buffer, reducing (6X)

Laemmli SDS sample buffer is used to prepare protein samples in the Laemmli SDS-PAGE system.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
50 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61337.AC
also known as J61337-AC
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Specifications
Concentration6X
For Use With (Application)Laemmli SDS-PAGE
FormLiquid
Quantity25 mL
ColorBlue
View more
For protein sample preparation to be used in the Laemmli SDS-PAGE system. Laemmli SDS sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Further, it is used for polyacrylamide protein gel analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • This compound is composed of several compounds, including 375 mM Tris-HCl (pH 6.8), 9% SDS, 50% glycerol, 9% β-mercaptoethanol, 0.03% bromophenol blue
  • The SDS compound present in the Laemmli SDS sample buffer binds noncovalently to proteins. SDS is negatively charged and can mask the intrinsic charge of the protein it binds

Application

  • Laemmli SDS sample buffer is used to denature proteins and for protein sample preparation to be used in the SDS-PAGE system
  • This sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of the running gel
  • It is also used for polyacrylamide protein gel analysis
RUO – Research Use Only