How many units of clostripain are in one milligram (dry weight) of Clostripain, Clostridium histolyticum?
Each mg of Clostripain, Clostridium histolyticum (dry weight) contains a minimum of 50.0 units of clostripain.
Search
View additional product information for Clostripain, Clostridium histolyticum, Thermo Scientific Chemicals - FAQs (J61362.MCR, J61362.MA)
2 product FAQs found