L-Histidine monohydrochloride monohydrate, Cell Culture Reagent
L-Histidine monohydrochloride monohydrate, Cell Culture Reagent
L-Histidine monohydrochloride monohydrate, Cell Culture Reagent
Thermo Scientific Chemicals

L-Histidine monohydrochloride monohydrate, Cell Culture Reagent

CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61465.09
also known as J61465-09
Price (USD)
24.20
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
24.20
Each
Chemical Identifiers
CAS5934-29-2
IUPAC Name(2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
Molecular FormulaC6H12ClN3O3
InChI KeyCMXXUDSWGMGYLZ-XRIGFGBMSA-N
SMILESO.Cl.N[C@@H](CC1=CNC=N1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay (special)Chloride: 16.6-17.1%
Impurity contentSulfate: ≤0.02%
pH3.5-4.5
Assay from Supplier's CofA≥98.0%
View more
L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

Solubility
Freely soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Thomas Osterberg.; Tommy Wadsten. Physical state of l-histidine after freeze-drying and long-term storage. European Journal of Pharmaceutical Sciences. 1999, 8, (4), 301-308.