SB 203580, 98%
SB 203580, 98%
SB 203580, 98%
Thermo Scientific Chemicals

SB 203580, 98%

Highly specific new inhibitor of p38 kinase and MAP Kinase homologues | CAS: 152121-47-6 | C21H16FN3OS, C21H16FN3OS | 377.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
50 mg
100 mg
Catalog number J61482.MC
also known as J61482-MC
Price (USD)
369.65
Online Exclusive
410.00
Save 40.35 (10%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
369.65
Online Exclusive
410.00
Save 40.35 (10%)
Each
Chemical Identifiers
CAS152121-47-6
IUPAC Name4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine
Molecular FormulaC21H16FN3OS
InChI KeyCDMGBJANTYXAIV-UHFFFAOYNA-N
SMILESCS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder or granular
Assay from Suppliers CofA≥97.5%
Appearance (Color)White to pale cream to pale yellow
Highly specific new inhibitor of p38 kinase and MAP Kinase homologues

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C