Phosphate, 0.5M buffer soln., pH 8.0
Phosphate, 0.5M buffer soln., pH 8.0
Phosphate, 0.5M buffer soln., pH 8.0
Thermo Scientific Chemicals

Phosphate, 0.5M buffer soln., pH 8.0

CAS: 7558-80-7119.976 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J61722.AK
also known as J61722-AK
Price (USD)
93.65
Special offer
Online exclusive
Ends: 28-Dec-2026
110.00
Save 16.35
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
93.65
Special offer
Online exclusive
Ends: 28-Dec-2026
110.00
Save 16.35
Each
Chemical Identifiers
CAS7558-80-7
IUPAC Namesodium dihydrogen phosphate
Molecular FormulaH2NaO4P
InChI KeyAJPJDKMHJJGVTQ-UHFFFAOYSA-M
SMILES[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH7.5-8.5
Concentration0.5M
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only