Thermo Scientific Chemicals

L-Tyrosine disodium salt dihydrate, Thermo Scientific Chemicals

Catalog number: J61770.A1
1 kg, Each
Thermo Scientific Chemicals

L-Tyrosine disodium salt dihydrate, Thermo Scientific Chemicals

Catalog number: J61770.A1
1 kg, Each
Quantity
Catalog number: J61770.A1
also known as J61770-A1
Price (USD)
Price: 477.00
Online price: 405.65
Your price:
Quantity
-

Chemical Identifiers

CAS
122666-87-9
IUPAC Name
disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate dihydrate
Molecular Formula
C9H13NNa2O5
InChI Key
WFTCFVUQORELJZ-CMIWSEPMNA-L
SMILES
O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O
Assay (special)
Endotoxin: ≤ 0.04 EU/mg
pH
10.5 to 11.5
Appearance (Color)
Pale cream to cream
Form
Powder
Heavy Metals
≤15 ppm (as Pb)

Description

Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text