Sulindac
Sulindac
Sulindac
Thermo Scientific Chemicals

Sulindac

CAS: 38194-50-2 | C20H17FO3S, C20H17FO3S | 356.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61772.06
also known as J61772-06
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.20
Save 11.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.20
Save 11.55
Each
Chemical Identifiers
CAS38194-50-2
IUPAC Name2-[(1Z)-5-fluoro-1-[(4-methanesulfinylphenyl)methylidene]-2-methyl-1H-inden-3-yl]acetic acid
Molecular FormulaC20H17FO3S
InChI KeyMLKXDPUZXIRXEP-MFOYZWKCNA-N
SMILESCC1=C(CC(O)=O)C2=CC(F)=CC=C2\C1=C/C1=CC=C(C=C1)S(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Purity≥97.0%
Appearance (Color)Yellow to orange
Sulindac inhibits the NF-κB pathway. Sulindac is a non-steroidal anti-inflammatory drug. Sulindac reduces the level and activity of several cyclin-dependent kinases and induces apoptosis. It is an antitumor agent. Sulindac is an inhibitor of Cox-1 and Cox-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Contents & Storage
-30°C to -10°C