Terfenadine
Terfenadine
Terfenadine
Thermo Scientific Chemicals

Terfenadine

CAS: 50679-08-8 | C32H41NO2
Have Questions?
Catalog NumberQuantity
J61936.MC
also known as J61936-MC
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61936.MC
also known as J61936-MC
Price (USD)
254.65
Special offer
Online exclusive
Ends: 28-Dec-2026
299.00
Save 44.35
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
254.65
Special offer
Online exclusive
Ends: 28-Dec-2026
299.00
Save 44.35
Each
Specifications
CAS50679-08-8
DescriptionWhite to Off-white
EINECS Number256-710-8
For Use With (Application)An antihistamine
FormPowder
View more
Terfenadine has been used to study the role of histamine in itch related to proteinase-activated receptors (PARs) in mice. Terfenadine has also been used to block histamine receptor type 1 to study the pathogenesis of 2,4-dinitrobenzene sulfonic acid (DNBS)-induced ulcerative colitis in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C