This product is animal origin and can only be shipped within the United States
Ribonuclease A, bovine pancreas, purified
Thermo Scientific Chemicals

Ribonuclease A, bovine pancreas, purified

Used to remove RNA from DNA plasmid preparations and protein samples. Ribonuclease A is used for RNase protection assays, to remove unspecifically bound RNA, analysis of RNA sequences, to hydrolyze RNA contained in protein samples, and the purification of DNA.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61996.MB
also known as J61996-MB
Price (USD)
-
Quantity:
25 mg
Request bulk or custom format
Chemical Identifiers
CAS9001-99-4
IUPAC NameRibonuclease A
MDL NumberMFCD00132181
SpecificationsSpecification SheetSpecification Sheet
AbsorbanceA280nm @ mg/ml solution: 0.49-0.66
Protease activity>3000 units/mgdw
Unit definitionOne unit hydrolyzes RNA from yeast, liberating soluble oligonucleotide and causing an increase in A260 of 1.0 at 37°C, pH 5.0. [One unit is equal to 50 Kunitz units]

Ribonuclease A is used to remove RNA from DNA plasmid preparations and protein samples. Ribonuclease A is used for RNase protection assays, to remove unspecifically bound RNA, analysis of RNA sequences, to hydrolyze RNA contained in protein samples, and the purification of DNA.

Ribonuclease A from bovine pancreas has been used in a study to assess hybridase activity of human ribonuclease-1. Ribonuclease A from bovine pancreas has also been used in a study to investigate particle-based and monolithic columns for cation exchange protein displacement chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only