5-Fluorouridine, 97%
5-Fluorouridine, 97%
5-Fluorouridine, 97%
Thermo Scientific Chemicals

5-Fluorouridine, 97%

CAS: 316-46-1 | C9H11FN2O6 | 262.193 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J62083.03
also known as J62083-03
Price (USD)
60.65
Online exclusive
67.20
Save 6.55
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
60.65
Online exclusive
67.20
Save 6.55
Each
Specifications
Beilstein33662
CAS316-46-1
ChEBICHEBI:40154
Chemical Name or Material5-Fluorouridine
ColorWhite
View more
5-Fluorouridine exhibits anticancer activity. It is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs. It is an active metabolite of Doxifluridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluorouridine exhibits anticancer activity. It is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs. It is an active metabolite of Doxifluridine.

Solubility
Soluble in Methanol (Slightly), Water (Slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Store at 4°C.
RUO – Research Use Only

General References:

  1. Ales Obrdlik; Piergiorgio Percipalle. The F-actin severing protein cofilin-1 is required for RNA polymerase II transcription elongation. Nucleus (Austin, Tex. : Print).2011, 2, (1), 72-79.
  2. P V Sahasrabudhe; R T Pon, W H Gmeiner. Effects of site-specific substitution of 5-fluorouridine on the stabilities of duplex DNA and RNA. Nucleic Acids Research.1995, 23, (19), 3916-3921.