Sulindac Sulfide
Sulindac Sulfide
Sulindac Sulfide
Thermo Scientific Chemicals

Sulindac Sulfide

CAS: 32004-67-4 | C20H17FO2S, C20H17FO2S | 340.41 g/mol
Have Questions?
Catalog NumberQuantity
J62104.MC
also known as J62104-MC
100 mg
Catalog number J62104.MC
also known as J62104-MC
Price (USD)
371.65
Online exclusive
413.00
Save 41.35
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
371.65
Online exclusive
413.00
Save 41.35
Each
Specifications
CAS32004-67-4
DescriptionYellow
EINECS Number250-892-2
For Use With (Application)Active metabolite of sulindac; inhibits cyclooxygenase-1
FormPowder
View more
It is also shown to inhibit growth and induce apoptosis in human prostate cancer cells through a COX-1 and COX-2 independent mechanism. Sulindac sulfide is an analgesic that has antiproliferative and apoptotic effects. It inhibits the expression and activity of cyclooxygenase-2 in human colon cancer cells, and reduces tumor burden in adenomatous polyposis patients. Human endothelial cells, HMEC-1 were treated with Sulindac sulfide and the effect on cell survival was studies by MTT assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only