Sodium cacodylate, 0.1M buffer soln., pH 6.8, Thermo Scientific Chemicals
Sodium cacodylate, 0.1M buffer soln., pH 6.8, Thermo Scientific Chemicals
Sodium cacodylate, 0.1M buffer soln., pH 6.8, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium cacodylate, 0.1M buffer soln., pH 6.8, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J62202.AE
also known as J62202-AE
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62202.AE
also known as J62202-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS6131-99-3
IUPAC Namesodium dimethylarsinate
Molecular FormulaC2H6AsNaO2
InChI KeyIHQKEDIOMGYHEB-UHFFFAOYSA-M
SMILES[Na+].C[As](C)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
pH6.6-7.0
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only