This product is animal origin and can only be shipped within the United States
Deoxyribonuclease I, bovine pancreas
Thermo Scientific Chemicals

Deoxyribonuclease I, bovine pancreas

Deoxyribonuclease I, CAS # 9003-98-9, also known as DNase I, is an endonuclease isolated from bovine pancreas used mainly to remove deoxyribonucleic acid (DNA) from protein samples.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62229.03
also known as J62229-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS9003-98-9
IUPAC NameDeoxyribonuclease
SMILES*
MDL NumberMFCD00130918
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (unspecified)A280: Report value
FormLyophilized powder
Assay (special)Endotoxin: Report value
Unit definitionOne unit causes an increase in absorbance at 260nm of 0.001 per minute at 25 degrees C, pH 5.0, when acting on highly polymerized DNA in the presence of magnesium and calcium
View more
Deoxyribonuclease I is used for the removal of DNA from protein samples. DNAse I is used to nick DNA as a first step to incorporate labeled bases into DNA. The enzyme from Sigma has been used during the isolation of plasma membrane vesicles from Neurospora crassa cell culture.1 It has also been used along with trypsin for the preparation of single cell suspension from rat testes. Deoxyribonuclease I from bovine pancreas has been used in a study to compare several procedures for reducing RNase contamination in preparations of DNase. Deoxyribonuclease I from bovine pancreas has also been used in a study to investigate the effect of the composition of sodium dodecyl sulfate preparations on the renaturation of enzymes after polyacrylamide gel electrophoresis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Deoxyribonuclease I is an endonuclease that acts on phosphodiester bonds adjacent to pyrimidines to produce polynucleotides with terminal 5′-phosphates. In the presence of Mg2+, this compound cleaves each strand of DNA independently

Application

  • In biomedical research, this compound is used to nick DNA as a first step to integrate labeled bases into DNA
  • It can be used to eliminate genomic DNA from RNA solutions before RT-PCR
  • It is also used during isolation and purification of plasma membrane vesicles from Neurospora crassa cell culture
  • It can decrease viscosity in cell culture lysates, providing better yields in primary cell isolation
  • It is also commonly used in combination with trypsin for the preparation of single-cell suspension from rats
  • Deoxyribonuclease I, bovine pancreas has been used in some studies in order to compare different methods for reducing RNase contamination in preparations of DNase
  • This compound can be applied in studies to investigate the impact of the composition of sodium dodecyl sulfate (SDS) preparations on the renaturation of enzymes after polyacrylamide gel electrophoresis
RUO – Research Use Only