This product is animal origin and can only be shipped within the United States
Ribonuclease A, bovine pancreas, chromatographically purified
Thermo Scientific Chemicals

Ribonuclease A, bovine pancreas, chromatographically purified

Used to remove RNA from DNA plasmid preparations and protein samples. Ribonuclease A is used for RNase protection assays, to remove unspecifically bound RNA, analysis of RNA sequences, to hydrolyze RNA contained in protein samples, and the purification of DNA.
Have Questions?
Change viewbuttonViewtableView
Quantity:
200 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62232.EX3
also known as J62232-!3
Price (USD)
-
Quantity:
200 mg
Request bulk or custom format
Chemical Identifiers
CAS9001-99-4
IUPAC NameRibonuclease A
MDL NumberMFCD00132181
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormLyophilized powder
Unit definitionOne unit hydrolyzes yeast RNA liberating soluble oligonucleotide causing an increase absorbance at 260nm of 1.0 at 37°C at pH 5.0 (Note: one Kunitz Unit equals 50 Worthington units)
units/mgdw≥2500 units per mg dry weight
UV/VisA280nm (@ 1 mg/ml): Report assay value
Ribonuclease A is used to remove RNA from DNA plasmid preparations and protein samples. Ribonuclease A is used for RNase protection assays, to remove unspecifically bound RNA, analysis of RNA sequences, to hydrolyze RNA contained in protein samples, and the purification of DNA. Ribonuclease A from bovine pancreas has been used in a study to assess hybridase activity of human ribonuclease-1. Ribonuclease A from bovine pancreas has also been used in a study to investigate particle-based and monolithic columns for cation exchange protein displacement chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only