This product is animal origin and can only be shipped within the United States
Collagenase, Type I, Clostridium histolyticum
Thermo Scientific Chemicals

Collagenase, Type I, Clostridium histolyticum

Type I Collagenase (CAS # 9001-12-1) is one of seven collagenases, enzymes responsible for collagen degradation.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62406.03
also known as J62406-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS9001-12-1
SpecificationsSpecification SheetSpecification Sheet
Unit definitionOne unit liberates one micromole of L-leucine equivalents from collagen in 5 hours at 37?C, pH 7.5
units/mgdwTryptic activity: ≤ 0.5
Appearance (Color)White to cream
Assay (special)Endotoxin (EU/mg dry wet): Report value
units/mgdwCaseinase: ≥ 200
View more
Collagenase, Type I, Clostridium histolyticum is used as a powder and solvent injection kit for the treatment of dupuytren's contracture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Type I collagenase is a collagen-specific enzyme required for the proper degradation of connective tissue
  • Type I collagenase is isolated from Clostridium histolyticum
  • It is implicated in connective tissue disorders such as Peyronie’s and Dupuytren’s

Application

  • Type I collagenase targets specific alpha helices within collagen
  • It is suitable for use in cell culture and biochemistry applications
RUO – Research Use Only

Frequently asked questions (FAQs)

How should I prepare Collagenase, Type I, Clostridium histolyticum (Cat. Nos. J62406.xx) for cell digestions?

For cell digestion, we recommend preparing collagenase using a salt solution such as Hank's Balanced Salt or Earle's Balanced Salt solution at concentrations in the range of 0.1-5 mg/mL. BSA (0.5%) or serum (5-10%) can be added to stabilize cells during digestion.

What is the procedure for using Collagenase, Type I, Clostridium histolyticum (Cat. No. J62406.xx) for assaying enzymatic activity?

For assaying enzymatic activity using Collagenase, Type I, Clostridium histolyticum (Cat. No. J62406.xx), we recommend the following procedure:

  1. Prepare enzyme stock solution. Create a stock solution by dissolving enzyme at a concentration of 1 mg/mL in 0.05 M TES with 0.36 mM calcium chloride, pH 7.5.
  2. Prepare enzyme working dilutions for assay. We recommend making either a 1/10 dilution or a 1/20 dilution of the stock solution above.
  3. Weigh 25 mg of Collagenase, Type I, Clostridium histolyticum into each of four test tubes. Include at least two tubes to serve as blanks. The blanks will not contain enzyme. Add 5.0 mL of 0.05 M TES buffer to the tubes (including the blanks) and incubate at 37°C for 15 minutes.
  4. Start the reaction by adding 0.1 mL of enzyme dilution to appropriate tubes. For blanks, substitute 0.1 mL of enzyme dilution with 0.1 mL of 0.05 M TES buffer.
  5. After 5 hours, stop the collagenase reaction by transferring 0.2 mL of solution from the experimental or blank test tubes to tubes containing 1.0 mL of ninhydrin-citric acid mixture. Make sure not to transfer over the collagen.
  6. Heat the test tubes for 20 minutes in a boiling water bath and then let them cool.
  7. After cooling, dilute with 5 mL of 50% n-propanol and let stand for 15 minutes.
  8. Read absorbance at 600 nm and use an L-leucine standard curve to determine micromoles amino acid equivalent to leucine liberated.

What storage conditions should I use for Collagenase, Type I, Clostridium histolyticum (Cat. No. J62406.xx)?

We recommend storing lyophilized powder at 2-8°C. Solutions should be stored at -20°C in aliquots to limit the number of freeze/thaw cycles. Freeze-thaw cycles will damage the enzyme solution.

What are the activators and inhibitors of Collagenase, Type I, Clostridium histolyticum (Cat. No. J62406.xx)?

Activators: Ca2+ and Zn2+ are collagenase activators.
Inhibitors: EDTA, EGTA, DTT, β-mercaptoethanol, o-phenanthroline and glutathione (reduced) are collagenase inhibitors.