Ionomycin, 96%
Ionomycin, 96%
Ionomycin, 96%
Thermo Scientific Chemicals

Ionomycin, 96%

Highly selective and potent calcium ionophore | CAS: 56092-81-0 | C41H72O9, C41H72O9 | 709.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number J62448.LB0
also known as J62448-#0
Price (USD)
342.65
Special offer
Online exclusive
Ends: 28-Dec-2026
403.00
Save 60.35
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
342.65
Special offer
Online exclusive
Ends: 28-Dec-2026
403.00
Save 60.35
Each
Chemical Identifiers
CAS56092-81-0
IUPAC Name(4R,6S,8S,10Z,12R,14R,16E,18R,19R,20S,21S)-11,19,21-trihydroxy-22-[(2S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9-oxodocosa-10,16-dienoic acid
Molecular FormulaC41H72O9
InChI KeyPGHMRUGBZOYCAA-OJFQOPKISA-N
SMILESC[C@H](CCC(O)=O)C[C@H](C)C[C@H](C)C(=O)\C=C(/O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)CC1CC[C@](C)(O1)C1CC[C@](C)(O1)[C@@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolution in ethanol
Mass SpecM-H+ : Report value (formula weight is 709 g/mol)
Assay from Supplier's CofA≥95% (HPLC)
Thin Layer Chromatography≥95.0% purity
Ionomycin free acid is a beneficial tool in research to raise intracellular calcium levels. Also this antibiotic can be used to study the calcium transport across the plasma membrane. It has been shown to induce central demyelination , inhibit adrenal bovine TREK-1 channels , and to regulate cell division of mature human B cells . It is used to study the effects of calcium flux on endoplasmic reticulum (ER) stress, mitochondrial stress and intrinsic apoptosis mechanisms. It is also used to stimulate the intracellular production of the cytokines, interferon, perforin, IL-2, and IL-4 usually in conjunction with PMA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C

Frequently asked questions (FAQs)

What is the concentration of the Ionomycin, 96% solution?

Ionomycin, 96% is a 10 mg/mL solution of ionomycin in ethanol.