Ionomycin, 96% - FAQs
Ionomycin, 96% - FAQs
View additional product information for Ionomycin, 96% - FAQs (J62448.MCR, J62448.LB0)
1 product FAQs found
What is the concentration of the Ionomycin, 96% solution?
Ionomycin, 96% is a 10 mg/mL solution of ionomycin in ethanol.