What are the concentrations of the components in RIPA buffer (5X) (Cat. No. J62524.xx)?
RIPA buffer (5X) contains 250 mM Tris-HCl (pH 7.4), 750 mM NaCl, 5% NP-40, 2.5% sodium deoxycholate, and 0.5% SDS.
Search
View additional product information for RIPA buffer (5X) - FAQs (J62524.AD, J62524.AE)
1 product FAQs found