Amrinone, 98%
Amrinone, 98%
Amrinone, 98%
Thermo Scientific Chemicals

Amrinone, 98%

Phosphodiesterase III inhibitor and inhibits platelet aggregation | CAS: 60719-84-8 | C10H9N3O
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62655.03
also known as J62655-03
Price (USD)
225.65
Online exclusive
251.00
Save 25.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
225.65
Online exclusive
251.00
Save 25.35
Each
Specifications
Assay98%
CAS60719-84-8
DescriptionYellow
EINECS Number262-390-0
For Use With (Application)Phosphodiesterase III inhibitor and inhibits platelet aggregation
View more
Phosphodiesterase III inhibitor and inhibits platelet aggregation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C