This product is animal origin and can only be shipped within the United States
Lipase, from porcine pancreas
Thermo Scientific Chemicals

Lipase, from porcine pancreas

 
Have Questions?
Catalog NumberQuantity
J62903.36
also known as J62903-36
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62903.36
also known as J62903-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS9001-62-1
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying5.0% max.
Assay (unspecified)Lipase: 24-50 USP units/mg
Lipase, is used as a catalyst in the preparation of biodiesels and in the preparation of esters of chiral epoxy alcohols. Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only