L-Valine, Cell Culture Reagent
L-Valine, Cell Culture Reagent
L-Valine, Cell Culture Reagent
Thermo Scientific Chemicals

L-Valine, Cell Culture Reagent

CAS: 72-18-4 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
1 kg
Catalog number J62943.06
also known as J62943-06
Price (USD)
29.20
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
29.20
Each
Chemical Identifiers
CAS72-18-4
IUPAC Name(2S)-2-amino-3-methylbutanoic acid
Molecular FormulaC5H11NO2
InChI KeyKZSNJWFQEVHDMF-BYPYZUCNSA-N
SMILESCC(C)[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Total microorganism: ≤ 100 CFU/gm; Mold: ≤ 10; Colliform: Negative
Residue on ignition≤ 0.10 %
Assay (unspecified)Endotoxin: ≤ 6.0
Heavy Metals≤ 10 ppm as lead
Loss on Drying≤ 0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only