This product is animal origin and can only be shipped within the United States
Superoxide Dismutase, bovine erythrocytes
Thermo Scientific Chemicals
Superoxide Dismutase, bovine erythrocytes
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 mg
10 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63003.MA
also known as J63003-MA
Price (USD)
-
Quantity:
10 mg
Request bulk or custom format
Chemical Identifiers
CAS9054-89-1
SpecificationsSpecification SheetSpecification Sheet
FormLyophilized powder
units/mgdw1,400 units per mg dry weight min.
Unit definitionOne Unit inhibits by 50% the maximum reduction of nitro blue tetrazolium under the specified conditions
UV/VisAbsorbance(280nm, 1mg/ml): Report Value
Superoxide dismutase from bovine erythrocytes has been used in a study to assess a kinetic model of radiation-induced inactivation of superoxide dismutase in nitrous oxide-saturated solutions. 1 Superoxide dismutase from bovine erythrocytes has also been used in a study to investigate the possible participation of superoxide anion in the intestinal tryptophan 2,3-dioxygenase reaction. It also catalyzes the dismutation of superoxide radicals to hydrogen peroxide and molecular oxygen. Plays a critical role in the defense of cells against the toxic effects of oxygen radicals. Competes with nitric oxide (NO) for superoxide anion (which reacts with NO to form peroxynitrite), thereby SOD promotes the activity of NO.3 SOD has also been shown to suppress apoptosis in cultured rat ovarian follicles, neural cell lines, and transgenic mice.