DNA Marker, high range, 1,000 Base Pair Ladder
Thermo Scientific Chemicals
DNA Marker, high range, 1,000 Base Pair Ladder
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
2 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63178.XF
also known as J63178-XF
Price (USD)
-
Quantity:
1 mL
Request bulk or custom format
Specifications
FormLiquid
Quantity1 mL
Storage Requirements−30° to −10°C
Product TypeMolecular Weight Marker
Unit SizeEach
DNA ladders are used in gel electrophoresis to determine the size and quantity of testing DNA fragments of genomic, plasmid, and PCR DNA. It is commonly used in the studies of plasmid DNA and PCR products. 1 kb DNA Ladder has been used as a molecular marker to determine the molecular weight and size of double stranded DNA during electrophoresis. 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb, 1 kb repeats from 3 to 6 kb, and 2 kb repeats from 6 to 10 kb. In NA electrophoresis, five ?l of the marker should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel. Suitable for use in Northern and Southern blotting. All DNA markers can be stained with ethidium bromide, SYBR™ Green, and Nancy-520.