TAPS, 0.2M buffer soln., pH 8.5, Thermo Scientific Chemicals
TAPS, 0.2M buffer soln., pH 8.5, Thermo Scientific Chemicals
TAPS, 0.2M buffer soln., pH 8.5, Thermo Scientific Chemicals
Thermo Scientific Chemicals

TAPS, 0.2M buffer soln., pH 8.5, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J63268.AE
also known as J63268-AE
100 mL
Catalog number J63268.AE
also known as J63268-AE
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Specifications
CAS91000-53-2
Recommended StorageAmbient Temperatures
Concentration0.2M buffer soln.
Molecular Weight (g/mol)265.256
Physical FormLiquid
View more
TAPS, is used in biochemistry and molecular biology. TAPS has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. TAPS has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TAPS, is used in biochemistry and molecular biology. TAPS has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. TAPS has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

Solubility
Soluble in water (333.33 mg/ml).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Julia Penso.; Rivka Beitner. Clotrimazole and bifonazole detach hexokinase from mitochondria of melanoma cells. European Journal of Pharmacology. 1998, 342, (1), 113-117.
  2. Maurizio Botta.; Federico Corelli.; Francesco Gasparrini.; Flavia Messina.; Claudia Mugnaini. Chiral Azole Derivatives. 4.1 Enantiomers of Bifonazole and Related Antifungal Agents: Synthesis, Configuration Assignment, and Biological Evaluation. J. Org. Chem. 2000, 65,(15), 4736-4739.