DL-Cystine
DL-Cystine
DL-Cystine
Thermo Scientific Chemicals

DL-Cystine

CAS: 923-32-0 | C6H12N2O4S2 | 240.292 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number J63564.06
also known as J63564-06
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.10
Save 9.45
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.10
Save 9.45
Each
Specifications
Beilstein1728095
CAS923-32-0
ChEBICHEBI:17376
Chemical Name or MaterialDL-Cystine
ColorWhite
View more
DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.

Solubility
Soluble in dilute acid and alkali solution, highly soluble in water, not soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Joseph Wells. Effect of water deprivation on uptake of dl-cystine-S35 in the hypothalamo-hypophysial system. Experimental Neurology. 1963, 8 (6), 470-481.
  2. Célia M C Faustino; António R T Calado; Luís Garcia-Rio; Célia M.C. Faustino; António R.T. Calado; Luís Garcia-Rio. Dimeric and monomeric surfactants derived from sulfur-containing amino acids. Journal of Colloid and Interface Science. 2010, 351 (2), 472-477.