Vinblastine Sulfate, 98%
Vinblastine Sulfate, 98%
Vinblastine Sulfate, 98%
Thermo Scientific Chemicals

Vinblastine Sulfate, 98%

Anticancer agent; microtubule disrupter; Induces apoptosis | CAS: 143-67-9 | C46H60N4O13S, C46H60N4O13S | 909.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63598.MF
also known as J63598-MF
Price (USD)
397.65
Special offer
Online exclusive
Ends: 30-Jun-2026
468.00
Save 70.35 (15%)
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
397.65
Special offer
Online exclusive
Ends: 30-Jun-2026
468.00
Save 70.35 (15%)
Each
Chemical Identifiers
CAS143-67-9
IUPAC Namemethyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
Molecular FormulaC46H60N4O13S
InChI KeyKDQAABAKXDWYSZ-JKDPCDLQSA-N
SMILESOS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale cream
FormCrystals or powder or granules or crystalline powder
Assay from Supplier's CofA≥97.5% (dry weight basis, India sourced material)
Assay from Supplier's CofA≥97.5% (HPLC, US sourced material)
CommentProduct sourced in US or India
Anticancer agent; microtubule disrupter; Induces apoptosis.Vinblastine sulfate is therapeutically used as an antineoplastic. It is also used in the treatment of Hodgkin disease, choriocarcinoma, acute and chronic leukemias.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Contents & Storage
2°C to 8°C