Temsirolimus, 99+%
Thermo Scientific Chemicals

Temsirolimus, 99+%

A Rapamycin derivative with anti-cancer activity that inhibits mTOR | CAS: 162635-04-3 | C56H87NO16
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
50 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63654.MC
also known as J63654-MC
Price (USD)
690.65
Special offer
Online exclusive
Ends: 30-Jun-2026
812.00
Save 121.35 (15%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
690.65
Special offer
Online exclusive
Ends: 30-Jun-2026
812.00
Save 121.35 (15%)
Each
Chemical Identifiers
CAS162635-04-3
IUPAC Name(1R,2R,4S)-4-[(2R)-2-[(1R,9S,12S,15R,18R,19R,21R,23S,30S,35R)-1,18-dihydroxy-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-2,3,10,14,20-pentaoxo-11,36-dioxa-4-azatricyclo[30.3.1.0⁴,⁹]hexatriaconta-16,24,26,28-tetraen-12-yl]propyl]-2-methoxycyclohexyl 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoate
Molecular FormulaC56H87NO16
InChI KeyCBPNZQVSJQDFBE-QYCDWSQESA-N
SMILES[H]C12CC[C@@H](C)[C@@](O)(O1)C(=O)C(=O)N1CCCC[C@@]1([H])C(=O)O[C@@]([H])(CC(=O)[C@H](C)C=C(C)[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)C=CC=CC=C(C)[C@H](C2)OC)[C@H](C)C[C@@H]1CC[C@@H](OC(=O)C(C)(CO)CO)[C@@H](C1)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream
Assay (HPLC)≥ 99.0 %
Assay by NMRConforms to structure
Temsirolimus is used as a rapamycin derivative with anti-cancer activity that inhibits mTOR. Temsirolimus is an antiproliferative and antiangiogenic, the first-in-class mTOR inhibitor approved for the treatment of patients with advanced poor prognosis renal cell carcinoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C