gamma-Secretase Inhibitor I
gamma-Secretase Inhibitor I
gamma-Secretase Inhibitor I
Thermo Scientific Chemicals

gamma-Secretase Inhibitor I

CAS: 133407-83-7 | C26H41N3O5, C26H41N3O5 | 475.63 g/mol
Have Questions?
Catalog NumberQuantity
J64068.LB0
also known as J64068-#0
5 mg
Catalog number J64068.LB0
also known as J64068-#0
Price (USD)
184.00
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
184.00
Each
Specifications
CAS133407-83-7
DescriptionWhite
For Use With (Application)Cell permeable. Inhibits →-secretase, a protease that cleaves the amyloid precursor protein (APP) to generate →-amyloid protein
FormSolid
Formula Weight475.62
View more
Cell permeable. Inhibits --secretase, a protease that cleaves the amyloid precursor protein (APP) to generate --amyloid protein

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C