1-beta-D-Arabinofuranosyluracil, 99%
1-beta-D-Arabinofuranosyluracil, 99%
1-beta-D-Arabinofuranosyluracil, 99%
Thermo Scientific Chemicals

1-beta-D-Arabinofuranosyluracil, 99%

CAS: 3083-77-0 | C9H12N2O6 | 244.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J64147.14
also known as J64147-14
Price (USD)
135.65
Special offer
Online exclusive
Ends: 30-Jun-2026
159.00
Save 23.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
135.65
Special offer
Online exclusive
Ends: 30-Jun-2026
159.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS3083-77-0
IUPAC Name1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H12N2O6
InChI KeyDRTQHJPVMGBUCF-CCXZUQQUSA-N
SMILESOC[C@H]1O[C@H]([C@@H](O)[C@@H]1O)N1C=CC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Water Content≤7.0%
Appearance (Color)White to off-white to pale yellow
Purity≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only