5-Azacytidine, 98%
5-Azacytidine, 98%
5-Azacytidine, 98%
Thermo Scientific Chemicals

5-Azacytidine, 98%

Causes DNA demethylation or hemi-demethylation; a powerful mutagen | CAS: 320-67-2 | C8H12N4O5 | 244.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J64159.06
also known as J64159-06
Price (USD)
275.65
Online Exclusive
306.00
Save 30.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
275.65
Online Exclusive
306.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS320-67-2
IUPAC Name4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydro-1,3,5-triazin-2-one
Molecular FormulaC8H12N4O5
InChI KeyNMUSYJAQQFHJEW-KVTDHHQDSA-N
SMILESNC1=NC(=O)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥97.5%
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
5-Azacytidine, its incorporation into RNA alters RNA synthesis and processing, and results in inhibition of protein synthesis. It has been used as a cancer chemotherapeutic agent. It is a powerful bacteriostatic, antitumor, and mutagenic agent; it also exhibits immunosuppressive, antimitotic, radioprotective, and virostatic effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Azacytidine, its incorporation into RNA alters RNA synthesis and processing, and results in inhibition of protein synthesis. It has been used as a cancer chemotherapeutic agent. It is a powerful bacteriostatic, antitumor, and mutagenic agent; it also exhibits immunosuppressive, antimitotic, radioprotective, and virostatic effects.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Siva P. Kumpatla.; Timothy C. Hall. Longevity of 5-azacytidine-mediated gene expression and re-establishment of silencing in transgenic rice. Plant Molecular Biology. 1998, 38, (6), 1113-1122.
  2. Sunil Rangappa.; Chen Fen.; Eng Hin Lee.; Ariff Bongso.; Eugene Sim Kwang Wei. Transformation of adult mesenchymal stem cells isolated from the fatty tissue into cardiomyocytes. The Annals of Thoracic Surgery. 2003, 75, (3), 775-779.