Cholesterol Esterase, from porcine pancreas
Thermo Scientific Chemicals
Cholesterol Esterase, from porcine pancreas
Have Questions?
Catalog NumberQuantity
J64282.EXR
also known as J64282-!R
100 U
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J64282.EXR
also known as J64282-!R
Price (USD)
-
Quantity:
100 U
Request bulk or custom format
Chemical Identifiers
CAS9026-00-0
SpecificationsSpecification SheetSpecification Sheet
Absorbanceat 280 nm (1 mg/mL): Report Value
CommentOne Unit hydrolyzes one micromole of cholesterol ester per minute at 37°C, pH 7.0
Protease≤80.0 ug/unit
units/mgdw≥300 units per gram dry weight
FormLyophilized Powder
Cholesterol Esterase is used for enzymatic determination of total cholesterol when coupled with cholesterol oxidize in clinical analysis. Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols.