3'-Amino-2',3'-dideoxythymidine, 99%
3'-Amino-2',3'-dideoxythymidine, 99%
3'-Amino-2',3'-dideoxythymidine, 99%
Thermo Scientific Chemicals

3'-Amino-2',3'-dideoxythymidine, 99%

CAS: 52450-18-7 | C10H15N3O4 | 241.25 g/mol
Have Questions?
Catalog NumberQuantity
J64342.03
also known as J64342-03
1 g
Catalog number J64342.03
also known as J64342-03
Price (USD)
80.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.40
Save 13.75 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.40
Save 13.75 (15%)
Each
Specifications
CAS52450-18-7
Chemical Name or Material3'-Amino-2',3'-dideoxythymidine
ColorWhite to Yellow
Formula Weight241.24
IUPAC Name1-[(2R,4S,5S)-4-amino-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only