5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt, 98%
5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt, 98%
5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt, 98%
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt, 98%

Substrate for beta-glucuronidase | CAS: 129541-41-9 | C14H12BrClNNaO7 | 444.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number J64360.MD
also known as J64360-MD
Price (USD)
192.65
Online Exclusive
214.00
Save 21.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
192.65
Online Exclusive
214.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS129541-41-9
IUPAC Namesodium 6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular FormulaC14H12BrClNNaO7
InChI KeyIBLSVGDGSKUDCT-UHFFFAOYNA-M
SMILES[Na+].OC1C(O)C(OC2=CNC3=CC=C(Br)C(Cl)=C23)OC(C1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Purity≥97.5% (HPLC)
Appearance (Color)White to off-white
FormPowder
Water Content≤10.0%
5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt is a chromogenic substrate for β-glucuronidase (GUS) gene detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-4-chloro-3-indolyl beta-D-glucuronide sodium salt is a chromogenic substrate for β-glucuronidase (GUS) gene detection.

Solubility
Soluble in water at 10mg/ml

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. H.Daniell. Foreign gene expression in chloroplasts of higher plants mediated by tungsten particle bombardment. Methods in Enzymology. 1993, 217,536-556.
  2. R.Bauer; F.Paltauf; S.D.Kohlwein. Functional expression of bacterial beta-glucuronidase and its use as a reporter system in the yeast Yarrowia lipolytica. Yeast. 1993, 9,(1), 71-75.