L-Cysteine methyl ester hydrochloride
L-Cysteine methyl ester hydrochloride
L-Cysteine methyl ester hydrochloride
Thermo Scientific Chemicals

L-Cysteine methyl ester hydrochloride

CAS: 18598-63-5 | C4H10ClNO2S | 171.639 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Catalog number J64436.30
also known as J64436-30
Price (USD)
314.65
Online Exclusive
349.00
Save 34.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
314.65
Online Exclusive
349.00
Save 34.35 (10%)
Each
Specifications
Beilstein3685824
CAS18598-63-5
Chemical Name or MaterialL-Cysteine methyl ester hydrochloride
ColorWhite
EINECS Number242-435-0
View more
L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.

Solubility
Soluble in water

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Leonidas Zervas.; Dimitrios M. Theodoropoulos. N-Tritylamino Acids and Peptides. A New Method of Peptide Synthesis1. J. Am. Chem. Soc. 1956, 78, (7), 1359-1363.
  2. Leonidas. Zervas.; Iphigenia. Photaki. On Cysteine and Cystine Peptides. I. New S-Protecting Groups for Cysteine. J. Am. Chem. Soc. 1962, 84, (20), 3887-3897.