8-Bromoadenosine-3',5'-cyclic monophosphate
8-Bromoadenosine-3',5'-cyclic monophosphate
8-Bromoadenosine-3',5'-cyclic monophosphate
Thermo Scientific Chemicals

8-Bromoadenosine-3',5'-cyclic monophosphate

CAS: 23583-48-4 | C10H11BrN5O6P | 408.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
250 mg
Catalog number J64469.MC
also known as J64469-MC
Price (USD)
315.65
Online Exclusive
351.00
Save 35.35 (10%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
315.65
Online Exclusive
351.00
Save 35.35 (10%)
Each
Specifications
CAS23583-48-4
ChEBICHEBI:64211
Chemical Name or Material8-Bromoadenosine-3',5'-cyclic monophosphate
ColorBrown
EINECS Number245-760-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only