2'-Amino-2'-deoxyguanosine, 98%
2'-Amino-2'-deoxyguanosine, 98%
2'-Amino-2'-deoxyguanosine, 98%
Thermo Scientific Chemicals

2'-Amino-2'-deoxyguanosine, 98%

CAS: 60966-26-9 | C10H14N6O4 | 282.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number J64650.MD
also known as J64650-MD
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
186.00
Save 27.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
186.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS60966-26-9
IUPAC Name2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular FormulaC10H14N6O4
InChI KeyROPTVRLUGSPXNH-SJLGBTOWNA-N
SMILESN[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5% (HPLC)
FormPowder
Appearance (Color)White to pale yellow to pale cream
2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Morio Ikehara.; Tokumi Maruyama.; Hiroko Miki. A new method for the synthesis of 2'-substituted purine nucleosides total synthesis of an antibiotic 2'-amino-2'-deoxyguanosine. Tetrahedron Letters. 1976, 17, (49), 4485-4488.
  2. Utagawa, T.; Morisawa, H.; Nakamatsu, T.; Yamazaki, A.; Yamanaka, S. Enzymatic synthesis of purine 2'-amino-2'-deoxyriboside. FEBS Letters. 1980, 119, (1), 101-104.