Calpain Inhibitor XII
Calpain Inhibitor XII
Calpain Inhibitor XII
Thermo Scientific Chemicals
Calpain Inhibitor XII
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number J64751.LB0
also known as J64751-#0
Price (USD)
412.65
Special offer
Online exclusive
Ends: 30-Jun-2026
485.00
Save 72.35 (15%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
412.65
Special offer
Online exclusive
Ends: 30-Jun-2026
485.00
Save 72.35 (15%)
Each
Specifications
DescriptionPale yellow
For Use With (Application)A cell-permeable dipeptidyl →-ketoamide compound that acts as a potent, selective, reversible, and active site inhibitor of calpain-1
FormSolid
Formula Weight482.60
Molecular FormulaC26H34N4O5
View more
A cell-permeable dipeptidyl --ketoamide compound that acts as a potent, selective, reversible, and active site inhibitor of calpain-1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C